Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kmt5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079347.1 | NP_001072815.1 | 3 - 4 | 414 | 89 | |
XM_018094658.1 | XP_017950147.1 | 2 - 3 | 362 | 89 | ||
GenBank mRNA | BC121601.1 | - | 3 - 4 | 414 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |