Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cox5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017049.3 | NP_001017049.1 | 2 - 3 | 317 | 97 | |
GenBank mRNA | BC135615.1 | - | 2 - 3 | 317 | 97 | |
CR761816.2 | - | 2 - 3 | 230 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |