Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017068.3 | NP_001017068.1 | 2 - 3 | 277 | 96 | |
XM_012957025.2 | XP_012812479.1 | 2 - 3 | 390 | 96 | ||
GenBank mRNA | BC135652.1 | - | 2 - 3 | 277 | 96 | |
CR761572.2 | - | 2 - 3 | 267 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |