Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033940.2 | NP_001029112.1 | 2 - 3 | 250 | 62 | |
XM_018097768.1 | XP_017953257.1 | 2 - 3 | 250 | 62 | ||
GenBank mRNA | AM051356.1 | - | 2 - 3 | 256 | 62 | |
BC135115.1 | - | 2 - 3 | 250 | 62 | ||
CR760359.2 | - | 2 - 3 | 230 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |