Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tubg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079041.1 | NP_001072509.1 | 1245 | 100 | ||
GenBank mRNA | BC121949.1 | - | 1245 | 100 |
Gene Symbol: LOC105945133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018097865.1 | XP_017953354.1 | 9 - 10 | 1193 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |