Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079061.1 | NP_001072529.1 | 4 - 5 | 781 | 100 | |
XM_018093095.1 | XP_017948584.1 | 5 - 6 | 715 | 100 | ||
GenBank mRNA | BC121984.1 | - | 4 - 5 | 781 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |