Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd5a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079071.1 | NP_001072539.1 | 1 - 2 | 382 | 74 | |
XM_012965419.2 | XP_012820873.1 | 2 - 3 | 509 | 74 | ||
XM_012965424.2 | XP_012820878.1 | 2 - 3 | 478 | 74 | ||
XM_012965426.2 | XP_012820880.1 | 2 - 3 | 511 | 74 | ||
GenBank mRNA | BC122039.1 | - | 1 - 2 | 382 | 74 | |
CU025196.1 | - | 2 - 3 | 479 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |