Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079079.1 | NP_001072547.1 | 3 - 4 | 271 | 73 | |
GenBank mRNA | BC122052.1 | - | 3 - 4 | 271 | 73 | |
CT030358.1 | - | 3 - 4 | 370 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |