Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: b3glct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079083.1 | NP_001072551.1 | 13 - 14 | 1239 | 109 | |
XM_012957278.2 | XP_012812732.1 | 12 - 13 | 1332 | 109 | ||
XM_018091349.1 | XP_017946838.1 | 11 - 12 | 1063 | 109 | ||
GenBank mRNA | BC122064.1 | - | 13 - 14 | 1239 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |