Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097181.1 | NP_001090650.1 | 3 - 4 | 616 | 61 | |
GenBank mRNA | BC127278.1 | - | 3 - 4 | 616 | 61 | |
CR761002.2 | - | 4 - 5 | 553 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |