Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097282.1 | NP_001090751.1 | 4 - 5 | 404 | 68 | |
XM_012971487.1 | XP_012826941.1 | 4 - 5 | 445 | 68 | ||
GenBank mRNA | EF010532.1 | - | 4 - 5 | 404 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis Probe spans exons |