Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097294.1 | NP_001090763.1 | 5 - 6 | 2133 | 64 | |
XM_012962531.2 | XP_012817985.1 | 5 - 6 | 2186 | 64 | ||
XM_012962532.2 | XP_012817986.1 | 5 - 6 | 2190 | 64 | ||
XM_018093725.1 | XP_017949214.1 | 5 - 6 | 1837 | 64 | ||
XM_018093726.1 | XP_017949215.1 | 5 - 6 | 2184 | 64 | ||
XM_018093727.1 | XP_017949216.1 | 5 - 6 | 2181 | 64 | ||
XM_018093728.1 | XP_017949217.1 | 5 - 6 | 1874 | 64 | ||
GenBank mRNA | BC129001.1 | - | 5 - 6 | 2133 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |