Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097318.1 | NP_001090787.1 | 7 - 8 | 1213 | 73 | |
XM_012960380.2 | XP_012815834.1 | 8 - 9 | 1033 | 73 | ||
XM_012960381.2 | XP_012815835.1 | 7 - 8 | 984 | 73 | ||
GenBank mRNA | BC135170.1 | - | 7 - 8 | 1213 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |