Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: heca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097357.1 | NP_001090826.1 | 3 - 4 | 1620 | 73 | |
GenBank mRNA | BC135464.1 | - | 3 - 4 | 1620 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |