Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097380.1 | NP_001090849.1 | 2 - 3 | 625 | 117 | |
GenBank mRNA | BC133720.1 | - | 2 - 3 | 625 | 117 | |
CR761154.2 | - | 2 - 3 | 390 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |