Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crebl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097383.1 | NP_001090852.1 | 3 - 4 | 613 | 80 | |
GenBank mRNA | BC135139.1 | - | 3 - 4 | 613 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |