Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esrrg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100210.1 | NP_001093680.1 | 6 - 7 | 1044 | 60 | |
XM_012962574.2 | XP_012818028.1 | 5 - 6 | 961 | 60 | ||
XM_012962575.2 | XP_012818029.1 | 4 - 5 | 859 | 60 | ||
XM_012962576.2 | XP_012818030.1 | 5 - 6 | 964 | 60 | ||
XM_012962577.2 | XP_012818031.1 | 4 - 5 | 882 | 60 | ||
XM_012962578.1 | XP_012818032.1 | 6 - 7 | 1044 | 60 | ||
XM_012962579.2 | XP_012818033.1 | 5 - 6 | 1012 | 60 | ||
XM_012962580.2 | XP_012818034.1 | 4 - 5 | 886 | 60 | ||
XM_018093742.1 | XP_017949231.1 | 5 - 6 | 937 | 60 | ||
XM_018093743.1 | XP_017949232.1 | 5 - 6 | 795 | 60 | ||
GenBank mRNA | BC136151.1 | - | 6 - 7 | 1044 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |