Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100224.1 | NP_001093694.1 | 1 - 2 | 431 | 72 | |
XM_012962582.2 | XP_012818036.1 | 1 - 2 | 671 | 72 | ||
GenBank mRNA | DQ675518.1 | - | 1 - 2 | 431 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |