Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prl.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100229.1 | NP_001093699.1 | 4 - 5 | 514 | 114 | |
GenBank mRNA | BC136077.1 | - | 4 - 5 | 514 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |