Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100247.1 | NP_001093717.1 | 8 - 9 | 2017 | 81 | |
GenBank mRNA | BC134803.1 | - | 8 - 9 | 2017 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |