Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmeff1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100268.3 | NP_001093738.2 | 8 - 9 | 1390 | 87 | |
XM_012964170.2 | XP_012819624.1 | 7 - 8 | 1374 | 87 | ||
GenBank mRNA | BC135422.2 | - | 8 - 9 | 1224 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |