Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102727.1 | NP_001096197.1 | 3 - 4 | 2427 | 82 | |
XM_012971500.2 | XP_012826954.1 | 3 - 4 | 2473 | 82 | ||
GenBank mRNA | BC135681.1 | - | 3 - 4 | 2427 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |