Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tti1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102727.1 | NP_001096197.1 | 4 - 5 | 2558 | 70 | |
XM_012971500.2 | XP_012826954.1 | 4 - 5 | 2604 | 70 | ||
GenBank mRNA | BC135681.1 | - | 4 - 5 | 2558 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |