Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102748.1 | NP_001096218.1 | 3 - 4 | 296 | 65 | |
GenBank mRNA | BC068216.1 | - | 3 - 4 | 296 | 65 | |
CR848327.2 | - | 3 - 4 | 290 | 65 | ||
CU025085.1 | - | 3 - 4 | 296 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |