Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dlc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102772.1 | NP_001096242.1 | 7 - 8 | 2254 | 90 | |
GenBank mRNA | BC135385.1 | - | 7 - 8 | 2254 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |