Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcakd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102776.1 | NP_001096246.1 | 3 - 4 | 450 | 76 | |
GenBank mRNA | BC135396.1 | - | 3 - 4 | 450 | 76 | |
CR761341.1 | - | 3 - 4 | 375 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |