Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102791.1 | NP_001096261.1 | 3 - 4 | 448 | 97 | |
XM_012955294.2 | XP_012810748.1 | 3 - 4 | 1811 | 97 | ||
GenBank mRNA | BC135453.1 | - | 3 - 4 | 448 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |