Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102833.1 | NP_001096303.1 | 12 - 13 | 1257 | 77 | |
GenBank mRNA | BC135644.1 | - | 12 - 13 | 1257 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |