Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nt5dc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102844.1 | NP_001096314.1 | 10 - 11 | 983 | 92 | |
XM_012958420.1 | XP_012813874.1 | 9 - 10 | 1137 | 92 | ||
XM_012958421.2 | XP_012813875.1 | 9 - 10 | 1099 | 92 | ||
GenBank mRNA | BC135684.1 | - | 10 - 11 | 983 | 92 | |
BC157658.1 | - | 9 - 10 | 880 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |