Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102857.1 | NP_001096327.1 | 3 - 4 | 490 | 87 | |
GenBank mRNA | BC135729.1 | - | 3 - 4 | 490 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |