Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pagr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102877.1 | NP_001096347.1 | 3 - 4 | 710 | 89 | |
XM_012954605.2 | XP_012810059.1 | 3 - 4 | 839 | 89 | ||
GenBank mRNA | BC135816.1 | - | 3 - 4 | 710 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |