Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c2orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102884.1 | NP_001096354.1 | 152 | 106 | ||
GenBank mRNA | BC135837.1 | - | 152 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis |