Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102942.1 | NP_001096412.1 | 11 - 12 | 1122 | 88 | |
XM_018093762.1 | XP_017949251.1 | 11 - 12 | 1155 | 88 | ||
GenBank mRNA | BC136067.1 | - | 11 - 12 | 1122 | 88 | |
CU075601.1 | - | 11 - 12 | 1087 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |