Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tppp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102996.1 | NP_001096466.1 | 3 - 4 | 586 | 102 | |
XM_012960482.2 | XP_012815936.1 | 4 - 5 | 705 | 102 | ||
XM_012960484.2 | XP_012815938.1 | 3 - 4 | 628 | 102 | ||
GenBank mRNA | BC136199.1 | - | 3 - 4 | 586 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |