Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: castor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103010.1 | NP_001096480.1 | 7 - 8 | 1070 | 77 | |
XM_018094494.1 | XP_017949983.1 | 7 - 8 | 1078 | 77 | ||
GenBank mRNA | BC136242.1 | - | 7 - 8 | 1070 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |