Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: g6pc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103061.1 | NP_001096531.1 | 3 - 4 | 497 | 66 | |
GenBank mRNA | BC135814.1 | - | 3 - 4 | 497 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |