Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crym | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103062.1 | NP_001096532.1 | 1 - 2 | 211 | 67 | |
XM_012969992.2 | XP_012825446.1 | 1 - 2 | 264 | 67 | ||
GenBank mRNA | BC135858.1 | - | 1 - 2 | 211 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |