Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c12orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016016.3 | NP_001016016.1 | 4 - 5 | 575 | 70 | |
GenBank mRNA | BC166293.1 | - | 4 - 5 | 575 | 70 | |
BC170669.1 | - | 4 - 5 | 899 | 70 | ||
BC170671.1 | - | 4 - 5 | 899 | 70 | ||
CR848210.2 | - | 4 - 5 | 920 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |