Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wnt9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103083.1 | NP_001096553.1 | 3 - 4 | 610 | 82 | |
GenBank mRNA | DQ658160.1 | - | 3 - 4 | 610 | 82 |
Gene Symbol: wnt9b.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002945312.4 | XP_002945358.3 | 2 - 3 | 640 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Xenopus tropicalis Probe spans exons |