Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110045.1 | NP_001103515.1 | 8 - 9 | 1435 | 92 | |
XM_018096093.1 | XP_017951582.1 | 9 - 10 | 1720 | 92 | ||
XM_018096094.1 | XP_017951583.1 | 8 - 9 | 1641 | 92 | ||
XM_018096095.1 | XP_017951584.1 | 7 - 8 | 1446 | 92 | ||
GenBank mRNA | BC153333.1 | - | 8 - 9 | 1435 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |