Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112961.2 | NP_001106432.1 | 2 - 3 | 221 | 100 | |
XM_012969733.2 | XP_012825187.1 | 2 - 3 | 440 | 100 | ||
GenBank mRNA | EL802545.1 | - | 2 - 3 | 245 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |