Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: awat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112975.1 | NP_001106446.1 | 5 - 6 | 484 | 92 | |
XM_012969998.2 | XP_012825452.1 | 5 - 6 | 1167 | 92 | ||
GenBank mRNA | BC153737.1 | - | 5 - 6 | 484 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |