Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pter | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113147.1 | NP_001106618.1 | 2 - 3 | 713 | 96 | |
XM_012964237.1 | XP_012819691.1 | 3 - 4 | 828 | 96 | ||
XM_012964238.1 | XP_012819692.1 | 3 - 4 | 796 | 96 | ||
XM_012964240.2 | XP_012819694.1 | 3 - 4 | 845 | 96 | ||
GenBank mRNA | BC155461.1 | - | 2 - 3 | 713 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |