Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113669.1 | NP_001107141.1 | 5 - 6 | 652 | 70 | |
XM_012960231.2 | XP_012815685.1 | 5 - 6 | 793 | 70 | ||
XM_012960256.2 | XP_012815710.1 | 5 - 6 | 765 | 70 | ||
GenBank mRNA | BC157660.1 | - | 5 - 6 | 652 | 70 | |
BC171317.1 | - | 4 - 5 | 506 | 70 | ||
BC171319.1 | - | 4 - 5 | 506 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |