Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113678.1 | NP_001107150.1 | 1 - 2 | 190 | 78 | |
GenBank mRNA | AM041994.1 | - | 1 - 2 | 190 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |