Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anos1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113688.1 | NP_001107160.1 | 4 - 5 | 716 | 78 | |
GenBank mRNA | BC155677.1 | - | 4 - 5 | 716 | 78 | |
BC170858.1 | - | 4 - 5 | 591 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |