Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: enkur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114042.1 | NP_001107514.1 | 1 - 2 | 121 | 71 | |
GenBank mRNA | BC157678.1 | - | 1 - 2 | 121 | 71 | |
BC171058.1 | - | 1 - 2 | 113 | 71 | ||
BC171060.1 | - | 1 - 2 | 113 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |