Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gnrhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114075.1 | NP_001107547.1 | 2 - 3 | 765 | 101 | |
GenBank mRNA | EF166086.1 | - | 2 - 3 | 765 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |