Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st6gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114199.1 | NP_001107671.1 | 5 - 6 | 1501 | 60 | |
XM_012956683.2 | XP_012812137.1 | 6 - 7 | 1770 | 60 | ||
XM_012956684.2 | XP_012812138.1 | 7 - 8 | 1828 | 60 | ||
XM_012956685.2 | XP_012812139.1 | 6 - 7 | 2395 | 60 | ||
GenBank mRNA | AJ627628.1 | - | 4 - 5 | 1291 | 60 | |
BC154086.1 | - | 5 - 6 | 1501 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |