Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114215.1 | NP_001107687.1 | 2 - 3 | 449 | 61 | |
GenBank mRNA | BC158266.1 | - | 2 - 3 | 449 | 61 | |
CR760594.2 | - | 2 - 3 | 500 | 61 | ||
CR942407.2 | - | 2 - 3 | 437 | 61 | ||
CT485738.1 | - | 2 - 3 | 503 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |